Learn More
Medchemexpress LLC Homatropine Bromide | 51-56-9 | 99.9% | 5 G

Supplier: Medchemexpress LLC HYB0547A5G
Homatropine Bromide is an orally active muscarinic acetylcholine receptor antagonist and functions as an anticholinergic agent. This compound is a white to off-white solid with a molecular weight of 356.25 and a chemical formula of C16H22BrNO3. It exhibits affinities for muscarinic receptors in the stomach and atria, and has been shown to block cardiovascular responses to vagal stimulation and acetylcholine in in vivo studies.
- Orally active muscarinic acetylcholine receptor antagonist
- Functions as an anticholinergic agent
- White to off-white solid
- Molecular weight: 356.25
- Chemical formula: C16H22BrNO3
- Soluble in DMSO and H2O at 100 mg/mL
- Solid storage: 4°C, sealed, away from moisture
- In solvent storage: -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.