Learn More
Apexbio Technology LLC Homatropine Methylbromide 80-49-9 50mg
SDP
Supplier:  Apexbio Technology LLC B160450
Encompass_Preferred
Homatropine Methylbromide (CAS 80-49-9) is a small molecule antagonist of muscarinic acetylcholine receptors (AChRs) It inhibits muscarinic signaling by blocking receptor activity on endothelial and smooth muscle cells as demonstrated in WKY-E and SHR-E models with IC50 values of 162 5 nM and 170 3 nM respectively This compound is utilized in studies investigating cholinergic modulation of vascular tone and smooth muscle function offering a tool for dissecting the roles of muscarinic receptors in cardiovascular and autonomic research contexts
Catalog No. 50-224-0811