Learn More
Medchemexpress LLC Homatropine methylbromide | 80-49-9 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB138850MG
Encompass_Preferred
Homatropine methylbromide is a muscarinic AChR antagonist that inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 values of 162.5 nM and 170.3 nM, respectively. It is intended for research use only and not for sale to patients.
  • Muscarinic AChR antagonist
  • Inhibits endothelial and smooth muscle muscarinic receptors
  • Available in solid and solution forms
Catalog No. 50-004-13519