Learn More
ABclonal Technology Homer1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2750220UL
Encompass_Preferred
This gene encodes a member of the homer family of dendritic proteins Members of this family regulate group 1 metabotrophic glutamate receptor function
Catalog No. 50-000-10040