Learn More
Medchemexpress LLC Homo Sildenafil | 642928-07-2 | 99.3% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY131100
Encompass_Preferred
Homo Sildenafil, an analog of Sildenafil, functions as a phosphodiesterase inhibitor. It is intended for research use only and is not sold to patients.
  • Acts as a phosphodiesterase inhibitor
  • Target is Phosphodiesterase (PDE)
  • Pathway classified under Metabolic Enzyme/Protease
Catalog No. 50-003-41771