Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Homo Sildenafil | 642928-07-2 | 99.3% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13110050MG

Encompass_Preferred

Homo Sildenafil is an analog of Sildenafil that acts as a phosphodiesterase inhibitor. This product is for research use only, not for sale to patients or for medical applications.

  • Molecular weight: 488.6
  • Molecular formula: C23H32N6O4S
  • Appearance: Solid, white to off-white
  • Target: Phosphodiesterase (PDE)
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility: 50 mg/mL in DMSO (in vitro)

Catalog No. 50-003-38603


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.