Learn More
TARGETMOL CHEMICALS INC Homoembelin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN42211MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Homoembelin (2,5-dihydroxy-3-nonyl-1,4-benzoquinone) is a pigment from E. ribes and C. monocarpus that has antibacterial effects and can be used to study multidrug-resistant bacterial infections.
Catalog No. 50-288-0245