Learn More
TARGETMOL CHEMICALS INC Homoeriodictyol 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN17401MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Homoeriodictyol, a naturally occurring, bitter-masking flavanone, as a promising agent to increase appetite and food intake. The flavanone homoeriodictyol can increase SGLT-1-mediated glucose uptake but decrease serotonin release in differentiated Caco-2 cells. Purity 99.96%
Catalog No. 50-287-5824