Learn More
Cole-Parmer HOMOGENIZER; LABGEN 850; 110V
Supplier:  Cole-Parmer 0472728
Encompass_Preferred
LabGEN 850 Homogenizer; 110V. Process virtually any laboratory sample in seconds Variable-speed motor performs from 5,000 to 25,000 rpmProcessing range from 200 uL to 5 LDigital, programmable speed and time controls
Catalog No. NC1871254