Learn More
Medchemexpress LLC Homogentisic acid lactone | 2688-48-4 | MFCD00005857 | 99.1% | 150.13 | C8H6O3 | 5 G

Supplier: Medchemexpress LLC HYW0174455G
Homogentisic acid lactone is a fungal metabolite supplied as a research-grade chemical reagent for biochemical and antifungal studies. It is commonly used in investigations of fungal pathogenicity and as a substrate in enzyme assays such as paraoxonase/arylesterase 1 (PON1) activity measurements. The material is provided with a certified purity and defined storage conditions for reliable experimental use.
- High purity suitable for analytical and biological research.
- Fungal metabolite with reported fungicidal activity.
- Usable as a substrate in PON1 and related enzyme assays.
- Molecular formula C8H6O3 and molecular weight 150.13.
- Storage recommendations provided for powder and solution forms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.