Learn More
Selleck Chemical LLC Homoharringtonine 1mg 26833-87-4 Omacetaxine mepesuccinate
SDP
Supplier:  Selleck Chemical LLC S90151MG
Encompass_Preferred
Homoharringtonine (Omacetaxine mepesuccinate), a plant alkaloid with antitumor properties, inhibits protein translation by preventing the initial elongation step of protein synthesis via an interaction with the ribosomal A-site. Homoharringtonine reversiblely inhibits IL-6-induced STAT3 Tyrosine 705 phosphorylation and reduced anti-apoptotic proteins expression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8357