Learn More
Medchemexpress LLC Homologous recombination-IN-1 | 391889-85-3 | 98.7% | 485.96 | 5 MG

Supplier: Medchemexpress LLC HY1481835MG
Homologous recombination-IN-1 is a novel RAD51-BRCA2 protein-protein interaction inhibitor (EC50=19 μM) that can interfere with homologous recombination. It reduces cell homologous recombination in a dose-dependent manner at concentrations of 10-40 μM over 5 hours.
- Purity: 98.65%
- Molecular weight: 485.96
- Formula: C28H24ClN3O3
- CAS No.: 391889-85-3
- Appearance: Solid
- Color: Light yellow to yellow
- Storage for powder: -20°C for 3 years; 4°C for 2 years
- Storage in solvent: -80°C for 6 months; -20°C for 1 month
- In vitro solubility: DMSO at 50 mg/mL (102.89 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.