Learn More
Cayman Chemical HomosIldenafIl 10mg
SDP
Supplier:  Cayman Chemical 3889110 MG
Encompass_Preferred
A PDE5 inhibitor (IC50 = 8.9 nM); has been found as an adulterant in dietary supplements for erectile dysfunction
Catalog No. 50-291-8866