Learn More
Medchemexpress LLC Homovanillic acid | 306-08-1 | 182.17 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN0384
Encompass_Preferred
Homovanillic acid (Vanilacetic acid) is a dopamine metabolite that has been associated with several conditions, including aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. It is intended for research use only and exhibits both antidepressant and neuroprotective activities.
  • Increases presynaptic membrane protein SYN1 in HT22 and rat hippocampal neurons, protecting synaptic structure.
  • Inhibits the degradation of autophagy marker proteins LC3 and SQSTM1/p62.
  • Reduces the formation of autophagosomes.
  • Exhibits antidepressant activities in mouse Corticosterone-induced depression models.
  • Exhibits neuroprotective activities in mouse Corticosterone-induced depression models.
  • Functions as an endogenous metabolite.
  • Serves as a disease marker for endocrine diseases, nervous system disorders, and immune system disorders.
Catalog No. 50-003-89454