Learn More
Medchemexpress LLC Homovanillyl alcohol | 2380-78-1 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN751310MM/1ML
Encompass_Preferred
Homovanillyl alcohol is a biological metabolite of Hydroxytyrosol, a phenolic compound naturally found in virgin olive oil and wine. This compound is recognized for its ability to protect red blood cells from oxidative injury and offers a protective effect against cardiovascular disease.
  • Biological metabolite of hydroxytyrosol
  • Protects red blood cells from oxidative injury
  • Exhibits protective effect against cardiovascular disease
  • Solid, white to off-white appearance
  • Classified as a phenol and monophenol
  • Derived from plants
Catalog No. 50-004-18795