Learn More
Medchemexpress LLC Honokiol | 35354-74-6 | 99.9% | 266.33 | 200 MG
Supplier: Medchemexpress LLC HYN0003200MG
Honokiol is a bioactive, biphenolic phytochemical that exhibits potent antioxidative, anti-inflammatory, antiangiogenic, and anticancer properties by targeting various signaling molecules. It is also known to inhibit the activation of Akt and can readily cross the blood-brain barrier.
- Potent antioxidative activity
- Anti-inflammatory activity
- Antiangiogenic activity
- Anticancer activity
- Inhibits Akt activation
- Readily crosses the blood-brain barrier