Learn More
Medchemexpress LLC Honokiol dichloroacetate | 1620160-42-0 | 99.1% | 488.19 g/mol | C22H18Cl4O4 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12429225MG
Encompass_Preferred
Honokiol DCA (honokiol dichloroacetate) is a dichloroacetate derivative of honokiol supplied as a high-purity research compound. It has been reported to inhibit the growth of human prostate cancer cells in vitro and to suppress androgen receptor protein levels. The material is a colorless to light yellow liquid with molecular formula C22H18Cl4O4 and molecular weight 488.19 g/mol. Recommended storage is solid at -20°C and in solvent at -80°C for longer-term storage.
Catalog No. 50-002-35241