Learn More
Grainger Hook and Loop 1 In PK100
Supplier:  Grainger 16U587
Encompass_Preferred
The 3M Scotchmate Reclosable Fastener SJ3541 is a hook and loop fastener strip that can be cut to size. It has adhesive backing for easy application and is made of nylon and polyester with acrylic adhesive. This VHB tape is 2 inches wide x 10 feet long and 0.045 inches thick. It has high shear and peel adhesion strength on most surfaces, with temperature resistance from -20°F to 200°F. The aggressive closure helps resist vibration and impact separation. This versatile fastener strip is ideal for mounting signs, trims, wire covers, and more.
Catalog No. 50-276-5046