Learn More
Grainger Hook and Loop 1 In PK100
Supplier:  Grainger 16U586
Encompass_Preferred
The Grainger 16U586 is a heavy duty industrial shelving unit made of steel with 5 shelves. It measures 36" W x 18" D x 72" H. The shelves are adjustable in 1" increments on the uprights. The weight capacity per shelf is 800 lbs. The shelves are made of steel with a black powder coat finish. The unit has 4 corner gussets for stability and 4 leveling feet. It can be used for industrial storage and organization. The shelving is easy to assemble and made in the USA.
Catalog No. 50-276-7661