Learn More
Grainger Hook and Loop 1x12 In Black PK50
Supplier:  Grainger 16U749
Encompass_Preferred
The Approved Vendor Hook and Loop Fastener Strip is a 1" wide strip of hook and loop fastener material that can be cut to desired length. It features a polyester hook side and nylon loop side with pressure sensitive adhesive backing for easy application. This versatile fastener strip can be used for a variety of purposes including crafts, DIY projects, wire management, and more. With a high operating temperature range of -20°F to 200°F, this hook and loop fastener is durable and reliable. The 10 foot long strip allows plenty of material for multiple uses.
Catalog No. 50-276-5245