Learn More
Grainger Hook and Loop 1x3 In Black PK100
Supplier:  Grainger 16U736
Encompass_Preferred
The 3M Scotchmate Reclosable Fastener SJ3541 is a hook and loop fastener strip that can be cut to desired length. It has adhesive backing for easy application and is 1" wide x 10 yards long. The hooks are on one side to adhere to the loops on the mating side. It can withstand temperatures up to 200°F and the adhesive withstands intermittent exposure to temperatures from -20°F to 200°F. This versatile fastener strip provides a quick, easy way to create a reclosable fastening system for a wide variety of applications.
Catalog No. 50-276-4896