Learn More
Grainger Hook and Loop 1x3 In Black PK200
Supplier:  Grainger 16U737
Encompass_Preferred
The 3M Hook and Loop Sanding Disc is a 5 inch diameter sanding disc with a hook and loop backing that attaches to hook and loop sanding tools. It features an open coat aluminum oxide mineral that cuts faster and cooler while resisting loading. The disc is designed for sanding wood, paint, metal and plastic. It has a maximum speed of 20,000 RPM and comes in grits ranging from 60 to 320. The discs can be used for both wet and dry sanding applications.
Catalog No. 50-276-5395