Learn More
Grainger Hook and Loop 1x3 In White PK100
Supplier:  Grainger 16U739
Encompass_Preferred
The 3M Scotchmate Reclosable Fastener SJ3522 Hook and Loop Fastener Strip is a 2" wide x 10 yard long roll of hook and loop fastener material. It features adhesive backing for easy application and a high cycle life hook and loop closure that allows for repeated fastening and unfastening. This versatile fastener strip can be cut to desired lengths and adhered onto surfaces to create a reusable closure. With a temperature resistance range of -20°F to 200°F, it is suitable for both indoor and outdoor use. The hook and loop strips securely adhere to each other, providing reliable closure strength.
Catalog No. 50-276-7051