Learn More
Grainger Hook Round 3/4 Dia Black PK1030
Supplier:  Grainger 16U757
Encompass_Preferred
The Grainger Round Hook is a 3/4" diameter black steel hook with a rounded tip. It has a working load limit of 75 lbs and is made in the USA. The hook has a zinc plating for moderate corrosion resistance. It can be used for a variety of light duty hanging and lifting applications. The hook comes in a pack of 10 and has an overall length of 4-1/2".
Catalog No. 50-276-7049