Learn More
Aviva Systems Biology Corp HOOK3 Antibody - C-terminal region (ARP65675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOOK3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human HOOK3. Alternate names: FLJ31058, HK3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-536