Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hordenine-d6 | 1346598-66-0 | 171.27 | C10H9D6NO | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0113S10MG

Encompass_Preferred

Hordenine-d6 is the deuterium-labeled analog of hordenine provided for use as an isotope-labeled standard or tracer in analytical, metabolic, and pharmacokinetic studies. It is intended for research applications where deuterated standards improve quantitation and metabolic tracking.

  • Deuterium-labeled hordenine for isotope tracing.
  • Molecular formula C10H9D6NO and molecular weight 171.27.
  • CAS number 1346598-66-0.
  • Recommended solubility: DMF, DMSO, ethanol at reported concentrations.
  • Storage: 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Supplied as a small research pack suitable for analytical workflows.

Catalog No. 50-002-96679


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.