Learn More
Bioworld Horse Blocker, 200 mL
Supplier:  Bioworld 209600022
Encompass_Preferred
Horse Blocker Blot blocking buffer used to block non-specific binding site for antibodies on the membrane. Technical Information: 10% Horse serum, 0.02% sodium azide in PBS
Catalog No. 50-198-544