Learn More
Rockland Immunochemicals Horse IgG F(ab')2 Antibody Peroxidase Conjugated 2 mg
SDP
Supplier:  Rockland Immunochemicals 6081304
Encompass_Preferred
Anti-Horse F(ab')2 peroxidase conjugated antibody generated in goat detects specifically horse IgG F(ab')2. This secondary peroxidase conjugated antibody anti-Horse is ideal for investigators who routinely perform titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3881