Learn More
Cusabio Technology LLC Horse Interleukin 6 IL-6ELISA
Supplier:  Cusabio Technology LLC CSBE16634HS
Encompass_Preferred
This kit detects IL-6 in equine serum plasma and tissue homogenates with a detection range of 3 12 pg/mL to 200 pg/mL and a sensitivity of 0 78 pg/mL Designed for quantitative analysis it employs a sandwich ELISA methodology requiring an incubation period of 1-5 hours and a sample volume of 50-100 L Absorbance measurements are performed at 450 nm ensuring accurate and reliable detection of IL-6 concentrations in horse-derived samples Suitable for research applications the assay provides high specificity and reproducibility for evaluating IL-6 levels in various biological matrices
Catalog No. 50-000-48256