Learn More
Grainger Hose Adapter 1-1/2 NPTF 1/4 NPTF
Supplier:  Grainger 14L770
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass fitting that converts 1-1/2" NPT female threads to 1/4" NPT female threads. It allows you to connect two hoses or pipes with different size thread connections. This adapter is made from brass for durability and resistance to corrosion. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. The dimensions are 2-1/2" length x 1-1/2" hex width. This hose adapter provides a reliable solution for joining mismatched thread sizes in fluid transfer applications.
Catalog No. 50-275-1652