Learn More
Grainger Hose Adapter 1-1/4 NPTF 5/8 NPTF
Supplier:  Grainger 14L777
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 1/2 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter allows you to connect a 3/4 in. ID hose to 1/2 in. NPT pipe fittings and valves. Made from brass for durability and corrosion resistance.
Catalog No. 50-274-9798