Learn More
Grainger Hose Adapter 1/2 NPTF 1-1/4 NPTF
Supplier:  Grainger 16P919
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/2" NPT male to 1/2" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +250°F. This adapter allows you to connect hydraulic hoses to pumps, valves, cylinders, and other components with 1/2" NPT threaded ports. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-4519