Learn More
Grainger Hose Adapter 1/2 NPTF 1/2 NPTF
Supplier:  Grainger 16P930
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/2" NPTF male to 1/2" NPTF female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect hydraulic hoses with different thread sizes. It is ideal for use in hydraulic systems in industrial equipment. The adapter measures 2-1/2" in length and 1-1/8" in diameter.
Catalog No. 50-276-4213