Learn More
Grainger Hose Adapter 1/2 NPTF 1/2 NPTF
Supplier:  Grainger 16P929
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/2" male JIC to 1/2" female JIC hydraulic hose adapter made of steel with a maximum pressure rating of 5,000 psi. This adapter allows you to connect two hydraulic hoses with different thread types. It has a working temperature range of -40°F to 250°F. The adapter is zinc plated for corrosion resistance. It meets or exceeds SAE J514 specifications. This adapter can be used in hydraulic applications to connect hoses with mismatched fittings.
Catalog No. 50-276-3803