Learn More
Grainger Hose Adapter 1/2 NPTF 1/4 NPTF
Supplier:  Grainger 16P905
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/4" hydraulic hose adapter made of steel with a zinc finish. It has a 37 degree JIC male end and a 37 degree JIC female end, allowing it to connect and adapt hydraulic hoses with 37 degree JIC fittings. This adapter can handle pressures up to 5000 PSI and temperatures from -40 to 250 degrees Fahrenheit. The adapter is 1-1/4" long with 1/4" NPT thread size. It meets or exceeds SAE J514 specifications. This hydraulic hose adapter provides a way to connect hydraulic hoses with different gendered 37 degree JIC fittings.
Catalog No. 50-276-4711