Learn More
Grainger Hose Adapter 1/2 NPTF 1/8 NPTF
Supplier:  Grainger 16P921
Encompass_Preferred
The Grainger 16P921 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.6/0.8 amps at full/half load. This electric motor is designed for general purpose applications.
Catalog No. 50-276-4148