Learn More
Grainger Hose Adapter 1/2 NPTF 3/8 NPTF
Supplier:  Grainger 16P926
Encompass_Preferred
The Grainger 1/2" NPT Male x 3/4" Garden Hose Thread Swivel Adapter is made of brass for durability. It allows you to connect a 1/2" NPT threaded pipe to a standard 3/4" garden hose. The swivel design prevents kinking and twisting of the hose. This adapter is ideal for connecting air compressors, pumps, and other equipment with 1/2" NPT outlets to a garden hose. It has a maximum pressure rating of 300 PSI and temperature range of -20°F to 250°F. The dimensions are 1-1/2" length x 1-1/16" width. This adapter provides a secure connection between pipe and hose.
Catalog No. 50-276-4047