Learn More
Grainger Hose Adapter 1/8 NPTF 18 mm
Supplier:  Grainger 16P947
Encompass_Preferred
The Grainger 16P947 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is thermally protected and rated for continuous duty applications. It can provide 0.56 kW of output power. The motor dimensions are 9-1/2" long x 7-3/16" wide x 8-7/16" high and it weighs 30 pounds. The 16P947 electric motor is manufactured in the USA.
Catalog No. 50-276-3869