Learn More
Grainger Hose Adapter 1/8 NPTF 33 mm
Supplier:  Grainger 16P953
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1 inch hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5000 psi and temperature range of -40 to 212 degrees Fahrenheit. This adapter connects a 1 inch SAE 100R2 type AT hose to 1 inch JIC 37 degree female pipe threads. It is manufactured in the USA by Adaptall. The adapter body length is 2.75 inches.
Catalog No. 50-276-4630