Learn More
Grainger Hose Adapter 1 NPTF 1/4 NPTF
Supplier:  Grainger 16P908
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/4" NPT male to 1/4" NPT female hydraulic hose adapter made of zinc-plated steel. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +212°F. This adapter allows you to connect 1/4" NPT threaded ports to 1/4" hydraulic hose. It is designed for use in hydraulic systems to adapt hose ends. The zinc plating helps prevent rust and corrosion. This adapter measures 1-1/2" long x 1" wide and weighs 0.05 lbs. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-4146