Learn More
Grainger Hose Adapter 1 NPTF 1/8 NPTF
Supplier:  Grainger 16P904
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1" NPTF x 1/8" NPTF hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and temperature range of -40°F to 250°F. This adapter allows you to connect 1" NPTF threaded ports to 1/8" NPTF hydraulic hose fittings. It can be used in hydraulic applications to adapt hose sizes. This adapter is manufactured in the USA by Adaptall.
Catalog No. 50-276-3942