Learn More
Grainger Hose Adapter 2 NPTF 14 mm
Supplier:  Grainger 16P958
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 2 NPTF (female) to 14 mm hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and temperature range of -40°F to +212°F. This adapter allows you to connect 14 mm hydraulic hose to 2 NPTF threaded ports. It is manufactured in the USA by Adaptall. The overall dimensions are 2-1/2" length x 1-1/16" width.
Catalog No. 50-276-3974