Learn More
Grainger Hose Adapter 3/4 NPTF 3/8 NPTF
Supplier:  Grainger 16P909
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 3/4 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter is made of forged brass for durability and has a hex body for easy tightening. It can be used for air, water, oil, natural gas, and inert gas applications. The dimensions are 1-1/2 in. length x 1-1/16 in. width. This adapter converts hose to pipe thread connections.
Catalog No. 50-276-4147