Learn More
Grainger Hose Adapter 3/4 NPTF 3/8 NPTF
Supplier:  Grainger 16P911
Encompass_Preferred
The Grainger 1/2" NPT Male x 1/2" NPT Female Brass Hose Adapter is a brass fitting that allows you to connect two hoses with different thread sizes. This adapter has a 1/2" NPT male thread on one end and a 1/2" NPT female thread on the other end. It is made from brass for durability and resistance to corrosion. The maximum pressure rating is up to 600 PSI and the maximum temperature rating is 250°F. This adapter can be used for hydraulic applications as well as connecting air hoses. It provides a secure threaded connection to prevent leaks. The dimensions are 1-1/2" length x 1-1/16" width. This brass hose adapter is ideal for adapting hoses with mismatched threads.
Catalog No. 50-276-3930