Learn More
Grainger PARKER Hydraulic Hose Adapter, 1/2
Supplier:  Grainger 52JF81
Encompass_Preferred
The Parker Hydraulic Hose Adapter 1/2 is a hydraulic hose adapter with a 1/2" NPTF male thread and a 1/2" 37° JIC female swivel thread. It is made from zinc-plated steel for corrosion resistance and has a maximum working pressure of 5000 psi. This adapter allows you to connect a 1/2" NPTF threaded component to a 1/2" 37° JIC hydraulic hose fitting. It can be used in a variety of hydraulic applications to adapt threaded ports to hose connections. The zinc plating provides corrosion resistance for use in harsh environments. This adapter is manufactured in the USA by Parker, a leading manufacturer of hydraulic components and systems.
Catalog No. NC2596494