Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Hose Barb Connector, Polypropylene, 5/8 in, Mini
Supplier:  Accuflow Systems PFMS625PPAF
Encompass_Preferred
Saint-Gobain Pure-Fit SIB mini hose barb connector in polypropylene for 5/8 in (15.9 mm) tubing. The Smooth Inner Bore (SIB) design provides a seamless transition between tubing and fittings, minimizing dead space and reducing the risk of fluid entrapment, making it suitable for aseptic bioprocessing and single-use assemblies.
  • Smooth inner bore construction reduces dead space and turbulence.
  • Polypropylene body compatible with common bioprocess fluids.
  • Designed for 5/8 in (15.9 mm) tubing, mini connector configuration.
  • Provides secure hose-barb connection for single-use assemblies.
  • Eliminates entrapment points to minimize bacterial growth risk.
Catalog No. NC2683167