Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD2204 Quick-Disconnect Hose Barb Insert; Valved, 1/4"" Tube ID, 1/8"" Flow Size, Acetal
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0636045
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.