Learn More
BURKLE INC Y-connector, PVDF, f. Ø 0.28-0.35 in. (7-9 mm), cylindrical nozzles
Supplier:  BURKLE INC 87120008
Encompass_Preferred
Cylindrical Y hose connector to connect hoses.
Catalog No. 50-000-26013