Learn More
Bioworld Hosta Multiplication Medium Stage II, 50 L
Supplier:  Bioworld 306301664
Encompass_Preferred
Hosta Multiplication Medium Stage IIContains Macro and Micronutrients as described by Murashige & Skoog (1962) and the Vitamins as described by Linsmaier and Skoog (1965)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 43.39 g/LPartially soluble in water
Catalog No. 50-254-910