Learn More
Hach Company Hot plate, 4x4, digital, 115V, North American plug. NO CE MARK.
Supplier:  Hach Company 2881400
Encompass
Easy-to-read digital temperature settings features a flashing warning (CAUTION! HOT TOP) when the surface exceeds 50°C. Low-profile design to improve safety. Reflective, white ceramic top is smooth, seamless and resistant to alkali/acid solutions. Detachable cord. Built-in support rod holder (order support rod separately). Operating temperature range: 5 to 540°C (41 to 1004°F).
Catalog No. 50-199-6762